missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-chlorothiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10141312
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10141312 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10141312 Supplier Thermo Scientific Maybridge Supplier No. GK01295DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 88105-17-3
Molecular Formula C6H5ClO2S
Molecular Weight (g/mol) 176.614
MDL Number MFCD00068135
InChI Key ZWKYYONTMGVPSP-UHFFFAOYSA-N
Synonym methyl3-chlorothiophene-2-carboxylate, methyl 3-chloro-2-thiophenecarboxylate, 3-chlorothiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 3-chloro-, methyl ester, 3-chloro-2-thiophenecarboxylic acid methyl ester, pubchem9775, acmc-20ai7j, rarechem al bf 1228, maybridge1_003830, buttpark 37\08-38
PubChem CID 2777566
IUPAC Name methyl 3-chlorothiophene-2-carboxylate
SMILES COC(=O)C1=C(C=CS1)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 176.62
Percent Purity 97%
Chemical Name or Material Methyl 3-chlorothiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.