missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-cyanobenzoate, 95%, Thermo Scientific™

Product Code. 10622015
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10622015 1 g Amber Glass Bottle 1g
10065152 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10622015 Supplier Thermo Scientific Maybridge Supplier No. RJC00610DA
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 13531-48-1
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00220156
InChI Key XPBHWSMZTSSEJE-UHFFFAOYSA-N
Synonym methyl m-cyanobenzoate, benzoic acid, 3-cyano-, methyl ester, 3-cyanobenzoic acid methyl ester, methyl-3-cyanobenzoate, methyl3-cyanobenzoate, pubchem21088, acmc-1burk, intermediates-zcf02042, ksc180c9j, 3-cn-c6h4-cooch3
PubChem CID 83554
IUPAC Name methyl 3-cyanobenzoate
SMILES COC(=O)C1=CC=CC(=C1)C#N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 161.16
Percent Purity 95%
Chemical Name or Material Methyle3-cyanobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.