missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-hydroxybenzo[b]thiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10643393
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10643393 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10643393 Supplier Thermo Scientific Maybridge Supplier No. BTB04875DA
 Restricted Item
Estimated Shipment: 03-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 13134-76-4
Molecular Formula C10H8O3S
Molecular Weight (g/mol) 208.23
MDL Number MFCD00067732
InChI Key JVUOTPTULVZUTL-KTKRTIGZSA-N
Synonym methyl 3-hydroxybenzo b thiophene-2-carboxylate, methyl 3-hydroxybenzo b thiophene-3-carboxylate, 2-hydroxy methoxy methylidene-1-benzothiophen-3-one, 2z-2-hydroxy methoxy methylidene-1-benzothiophen-3-one, benzo b thiophene-2-carboxylicacid, 3-hydroxy-, methyl ester, benzo b thiophene-2-carboxylic acid, 3-hydroxy-, methyl ester, maybridge4_000295, methyl 3-hydroxy-benzothiophene2-carboxylate
PubChem CID 83165
SMILES CO\C(O)=C1/SC2=CC=CC=C2C1=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name methyl 3-hydroxy-1-benzothiophene-2-carboxylate
Formula Weight 208.24
Percent Purity 97%
Chemical Name or Material Methyl 3-hydroxybenzo[b]thiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.