missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10263232
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Amber Glass Bottle
Unit Size:
100g
10g
50g
This item is not returnable. View return policy

Product Code. 10263232

Brand: Thermo Scientific Maybridge SB01952EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

Molecular Formula C6H6O3S
MDL Number MFCD00055642

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 158.18
Percent Purity 97%
Chemical Name or Material Methyle3-hydroxythiophene-2-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.