missing translation for 'onlineSavingsMsg'
Learn More

Methyle4-methoxythiophene-3-carboxylate, 97%, Thermo Scientific™

Product Code. 10101312
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10101312 250 mg 250mg
10019551 1 g 1g
10337114 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10101312 Supplier Thermo Scientific Maybridge Supplier No. KM00444CB
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 65369-22-4
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.198
MDL Number MFCD00067995
InChI Key LQIVVUILGDRRHE-UHFFFAOYSA-N
Synonym methyl 4-methoxy-3-thiophenecarboxylate, 4-methoxy-thiophene-3-carboxylic acid methyl ester, 3-thiophenecarboxylic acid, 4-methoxy-, methyl ester, maybridge1_004625, ksc495s3h, methyl 4-methoxythiophene-3-arboxylate, 3-methoxythiophene-4-carboxylic acid methyl ester, 3-thiophenecarboxylicacid,4-methoxy-,methyl ester, 4-methoxy-3-thiophenecarboxylic acid methyl ester, 3-thiophenecarboxylicacid, 4-methoxy-, methyl ester
PubChem CID 2778377
IUPAC Name methyl 4-methoxythiophene-3-carboxylate
SMILES COC1=CSC=C1C(=O)OC

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 172.2
Percent Purity 97%
Chemical Name or Material Methyl 4-methoxythiophene-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.