missing translation for 'onlineSavingsMsg'
Learn More

Methyle5-(chlorosulfonyl)-4-methyl-2-thiophenecarboxylate, 97%, Thermo Scientific™

Product Code. 10682675
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10682675 5 g Amber Glass Bottle 5g
10252792 250 mg Amber Glass Bottle 250mg
10304202 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10682675 Supplier Thermo Scientific Maybridge Supplier No. MO00640DE
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 423768-46-1
Molecular Formula C7H7ClO4S2
Molecular Weight (g/mol) 254.699
MDL Number MFCD03407367
InChI Key YWBXGHFHMHUSKH-UHFFFAOYSA-N
Synonym methyl 5-chlorosulfonyl-4-methylthiophene-2-carboxylate, methyl 5-chlorosulfonyl-4-methyl-2-thiophenecarboxylate, 2-thiophenecarboxylicacid, 5-chlorosulfonyl-4-methyl-, methyl ester
PubChem CID 2779815
IUPAC Name methyl 5-chlorosulfonyl-4-methylthiophene-2-carboxylate
SMILES CC1=C(SC(=C1)C(=O)OC)S(=O)(=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 254.71
Percent Purity 97%
Chemical Name or Material Methyl 5-(chlorosulfonyl)-4-methyl-2-thiophenecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.