missing translation for 'onlineSavingsMsg'
Learn More

Methyle6-(1-pyrrolidinyl)nicotinate, 97%, Thermo Scientific™

Product Code. 10337074
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10337074 5 g 5g
10055262 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10337074 Supplier Thermo Scientific Maybridge Supplier No. MO07009DE
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 210963-94-3
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.245
MDL Number MFCD05664433
InChI Key OWOKTZZFJFFUKY-UHFFFAOYSA-N
Synonym methyl 6-pyrrolidinonicotinate, methyl 6-1-pyrrolidinyl nicotinate, methyl 6-pyrrolidin-1-yl nicotinate, methyl 6-pyrrolidin-1-yl pyridine-3-carboxylate, acmc-1cher, methyl 6-pyrrolidine-1-yl nicotinate, methyl 6-pyrrolidine-1-yl nicotinoate, methyl 6-pyrrolidin-1-ylnicotinate, methyl 6-pyrrolidinylpyridine-3-carboxylate, methyl 6-pyrrolidin-1-yl nicotinate;ethyl 6-pyrrolidin-1-yl pyridine-3-carboxylate
PubChem CID 2794748
IUPAC Name methyl 6-pyrrolidin-1-ylpyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)N2CCCC2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material Methyle6-(1-pyrrolidinyl)nicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.