missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue, Fisher BioReagents™

Product Code. 10307033
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10307033 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10307033 Supplier Fisher BioReagents Supplier No. BP117100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 190°C
CAS 7220-79-3
Extinction Coefficient 2000g(-1) cm(-1) min.
Molecular Formula C16H24ClN3O3S
MDL Number MFCD00150008
Merck Index 15, 6132
Solubility Information Solubility in water: slight
Identification Pass Test
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate
Molecular Weight (g/mol) 373.90
PubChem CID 104827
Lambda Max 660 to 664nm
Formula Weight 373.9
Loss on Drying 8.0 to 18.0%
Packaging Glass Bottle
Vapor Pressure Vapor Pressure: Negligible
Decomposition Information 190°C
Density 1.230g/cm³
Color Index No. 52015
Color Green
pH 3 to 4.5
Quantity 100 g
Physical Form Solid
Chemical Name or Material Methylene Blue
Show More Show Less
compliance-icons
Product Identifier
  • Methylene Blue
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.