missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue hydrate, 96+%, high purity biological stain

Product Code. 10004053 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10004053 1 kg Glass bottle 1kg
10741891 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10004053 Supplier Thermo Scientific Chemicals Supplier No. 229800010
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 122965-43-9 | C16H18ClN3S | 319.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122965-43-9
Molecular Formula C16H18ClN3S
Molecular Weight (g/mol) 319.85
MDL Number MFCD00150006
InChI Key CXKWCBBOMKCUKX-UHFFFAOYSA-M
Synonym Basic Blue 9 hydrate, C.I. 52015 hydrate
PubChem CID 16211647
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C

Specifications

Melting Point 100.0°C to 110.0°C
Dye Content 96.0 to 103.0% (on dried substance)
Infrared Spectrum Authentic
Loss on Drying 8.0% to 22.0% (105°C)
Packaging Glass bottle
Assay Percent Range 96+%
Color Green
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol
Quantity 1 kg
Physical Form Crystalline Powder
Formula Weight 319.85
Percent Purity ≥96%
Chemical Name or Material Methylene Blue hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.