missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue trihydrate, 95%, pure

Product Code. 10173030 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10173030 100 g Glass bottle 100g
10695172 250 g Glass bottle 250g
10175253 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10173030 Supplier Thermo Scientific Chemicals Supplier No. 126751000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7220-79-3 | C16H24ClN3O3S | 373.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7220-79-3
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
Synonym Basic Blue 9 trihydrate, C.I. 52015 trihydrate
PubChem CID 104827
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

Melting Point 100.0°C to 110.0°C
Infrared Spectrum Authentic
Loss on Drying 15% max. (105°C)
Packaging Glass bottle
Assay Percent Range 95%
Beilstein 27, 393
Merck Index 15, 6132
Color Green
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, insoluble in ether
Quantity 100 g
Physical Form Crystalline Powder With Metal Luster
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate
Formula Weight 373.89
Percent Purity 95%
Grade Pure
Chemical Name or Material Methylene Blue trihydrate, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Methylene Blue trihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Methylene blue trihydrate, Methylene blue

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.