missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue trihydrate, 95%, pure

Product Code. 10175253 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
250g
This item is not returnable. View return policy

Product Code. 10175253

Brand: Thermo Scientific Chemicals 126750010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7220-79-3 | C16H24ClN3O3S | 373.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7220-79-3
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
Synonym Basic Blue 9 trihydrate, C.I. 52015 trihydrate
PubChem CID 104827
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

Melting Point 100.0°C to 110.0°C
Assay Percent Range 95%
Beilstein 27, 393
Merck Index 15, 6132
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, insoluble in ether
Infrared Spectrum Authentic
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate
Formula Weight 373.89
Loss on Drying 15% max. (105°C)
Percent Purity 95%
Grade Pure
Packaging Glass bottle
Color Green
Quantity 1 kg
Physical Form Crystalline Powder With Metal Luster
Chemical Name or Material Methylene Blue trihydrate, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.