missing translation for 'onlineSavingsMsg'
Learn More

Methylene Chloride (Pentene Preservative), Fisher BioReagents

Product Code. 10512965
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10512965 4 L Glass Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10512965 Supplier Fisher BioReagents Supplier No. BP11864
  • ONLINE EXCLUSIVE PRICE
    187.00 €
    184.10 € / 4 L
    Save 2.90 €
    1% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This solvent is often used in automated synthesis procedures.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -97°C
Absorbance 1 max. at 233nm, 0.12 max. at 240nm, 0.01 max. at 254nm
CAS 110-83-8, 513-35-9, 67-56-1, 75-09-2
Color Colorless
Density 1.33g/cm³
Quantity 4 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4200 to 1.4230 (at 25°C)
Residue after Evaporation 1ppm max.
Assay Percent Range ≥99.9 %
Titratable Acid 0.0003mEq/g max.
Molecular Formula C6H10
Vapor Pressure 20mmHg at 3502°C
UN Number 1593
Water 25ppm max.
Merck Index 15, 6135
Synonym tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene
Boiling Point 39°C
InChI Key HGCIXCUEYOPUTN-UHFFFAOYSA-N
SMILES C1CCC=CC1
IUPAC Name cyclohexene
Molecular Weight (g/mol) 82.146
PubChem CID 8079
ChEBI CHEBI:36404
Formula Weight 84.93
Percent Purity ≥99.9%
Chemical Name or Material Methylene Chloride, Pentene Preservative
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methylene Chloride
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.