missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ α,β-Methyleneadenosine 5'-triphosphate trisodium salt

Product Code. 18084552
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18084552 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18084552 Supplier R&D Systems™ Supplier No. 3209/10
  • ONLINE EXCLUSIVE PRICE
    327.40 €
    294.00 € / 10mg
    Save 33.40 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Non-selective P2 agonist

P2-purinoceptor agonist. Also shown to inhibit adenylate cyclase (Ki = 0.5 mM).
TRUSTED_SUSTAINABILITY

Specifications

CAS 1343364-54-4
Quantity 10 mg
Product Line Tocris
Molecular Formula C11H15N5Na3O12P3
Formula Weight 571.15
Purity >98%
Molecular Weight (g/mol) 571.155
Chemical Name or Material α, β-Methyleneadenosine 5'-triphosphate trisodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.