missing translation for 'onlineSavingsMsg'
Learn More

Methylenecyclohexane, 98%

Product Code. 10135520 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10135520

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1192-37-6 | C7H12 | 96.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192-37-6
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
MDL Number MFCD00001661
InChI Key YULMNMJFAZWLLN-UHFFFAOYSA-N
Synonym methylenecyclohexane, cyclohexane, methylene, 1-methylenecyclohexane, methylene-cyclohexane, cyclohexylmethyl, methylcyclohexyl, acmc-209a1w, methylene cyclohexane
PubChem CID 14502
IUPAC Name methylidenecyclohexane
SMILES C=C1CCCCC1

Specifications

Color Colorless
Density 0.8000g/mL
Boiling Point 102.0°C to 103.0°C
Flash Point −006°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H10(=CH2)
Refractive Index 1.4465 to 1.4505
Quantity 1 g
Beilstein 05, 69
Specific Gravity 0.8
Formula Weight 96.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methylenecyclohexane
Show More Show Less
compliance-icons
Product Identifier
  • Methylenecyclohexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.