missing translation for 'onlineSavingsMsg'
Learn More

Methylenediphosphonic acid, 98%, Thermo Scientific Chemicals

Product Code. 15264684 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15264684 5 g Glass Bottle 5g
10562231 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15264684 Supplier Thermo Scientific Acros Supplier No. 215070050
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1984-15-2
Molecular Formula CH6O6P2
Molecular Weight (g/mol) 176.00
MDL Number MFCD00002139
InChI Key MBKDYNNUVRNNRF-UHFFFAOYSA-N
Synonym methylenediphosphonic acid, medronic acid, acido medronico, methylene diphosphonate, acide medronique, acidum medronicum, phosphonic acid, methylenebis, methylenebis phosphonic acid, mdp-bracco, methanediphosphonic acid
PubChem CID 16124
ChEBI CHEBI:43945
SMILES OP(O)(=O)CP(O)(O)=O

Specifications

Melting Point 200.0°C to 204.0°C
Color Blue
Boiling Point 82.0°C (6.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CH2[P(O)(OH)2]2
Quantity 5 g
UN Number 3261
Beilstein 01,IV,3071
Merck Index 15,5863
Solubility Information Solubility in water: soluble
IUPAC Name (phosphonomethyl)phosphonic acid
Formula Weight 176
Percent Purity 98%
Physical Form Moist Crystals or Crystalline Powder
Chemical Name or Material Methylenediphosphonic Acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Methylenediphosphonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.