missing translation for 'onlineSavingsMsg'
Learn More

Methylequinoxaline-6-carboxylate, 97%, Thermo Scientific™

Product Code. 10632833
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10632833 10 g Amber Glass Bottle 10g
10121172 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10632833 Supplier Thermo Scientific Maybridge Supplier No. RDP00059EA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 23088-23-5
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD00102618
InChI Key VFPWJISMXACHIG-UHFFFAOYSA-N
Synonym methyl 6-quinoxalinecarboxylate, 6-quinoxalinecarboxylic acid, methyl ester, maybridge1_006081, acmc-1cnh9, methyl6-quinoxalinecarboxylate, methyl quinoxaline-7-carboxylate, 6-quinoxalinecarboxylic acid methyl ester, 6-quinoxalinecarboxylicacid, methyl ester, 6-quinoxalinecarboxylic acid methyl ester;
PubChem CID 2781239
IUPAC Name methyl quinoxaline-6-carboxylate
SMILES COC(=O)C1=CC=C2N=CC=NC2=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material Methyl quinoxaline-6-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.