missing translation for 'onlineSavingsMsg'
Learn More

Methylgermanium trichloride, 97%

Product Code. 11304178
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
This item is not returnable. View return policy

Product Code. 11304178

Brand: Thermo Scientific Alfa Aesar 041449.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 993-10-2 | CH9Cl3Ge | 200.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylgermanium trichloride is used as a precursor for the synthesis of five-coordinated spirocyclic anionic methylgermanates by reacting with catechol. It is also involved in the photo-chemical adsorption of organo-germanium on a hydrogen terminated atomically flat silicon surface using xenon excimer light.

Solubility
Miscible with ether.

Notes
Sensitive to moisture and reacts violently with water. Also hydrolysis takes place when contact with water. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 993-10-2
Molecular Formula CH9Cl3Ge
Molecular Weight (g/mol) 200.06
MDL Number MFCD00013585
InChI Key NNMJXXWNUALEKD-UHFFFAOYSA-N
Synonym methyltrichlorogermane, trichloro methyl germane, methylgermanium trichloride, germanium methyl trichloride, germane, trichloromethyl, fefxfmqvsdtspa-uhfffaoysa, inchi=1/ch3cl3ge/c1-5 2,3 4/h1h3
PubChem CID 70436
IUPAC Name trichloro(methyl)germane
SMILES Cl.Cl.Cl.C[GeH3]

Specifications

Color Colorless to Yellow
Density 1.706 g/mL
Boiling Point 111°C
Flash Point 10°C (50°F)
Odor Chlorine-like
Linear Formula CH3GeCl3
Refractive Index 1.4685
Quantity 10 g
UN Number UN2924
Sensitivity Moisture sensitive
Formula Weight 193.99
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methylgermanium trichloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.