missing translation for 'onlineSavingsMsg'
Learn More

Methylguanidine hydrochloride, 98%

Product Code. 15452879
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452879 5 g 5g
15462879 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15452879 Supplier Thermo Scientific Alfa Aesar Supplier No. J60033.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22661-87-6 | C2H8ClN3 | 109.557 g/mol

Methylguanidine hydrochloride can be used in the synthesis of biaryl derivatives which were evaluated as BACE1 inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylguanidine hydrochloride can be used in the synthesis of biaryl derivatives which were evaluated as BACE1 inhibitors.

Solubility
Soluble in water (50 mg/ml - clear, colorless to light yellow solution).

Notes
Hygroscopic, store away form water/moisture. Store in freezer -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent and reducing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22661-87-6
Molecular Formula C2H8ClN3
Molecular Weight (g/mol) 109.557
MDL Number MFCD00012576
InChI Key VJQCNCOGZPSOQZ-UHFFFAOYSA-N
Synonym 1-methylguanidine hydrochloride, methylguanidine hydrochloride, methylguanidine hcl, n-methylguanidine hydrochloride, 1-methylguanidine hcl, guanidine, methyl-, hydrochloride, 2-methylguanidine hydrochloride, 1-methylguanidine hydrochloride 1:x, guanidine, methyl-, monohydrochloride, methyl guanidine hcl
PubChem CID 146724
IUPAC Name 2-methylguanidine;hydrochloride
SMILES CN=C(N)N.Cl

Specifications

Color White
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water (50mg/ml- clear,colorless to light yellow solution).
Formula Weight 109.56
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methylguanidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.