missing translation for 'onlineSavingsMsg'
Learn More

Methylguanidine hydrochloride, Thermo Scientific™

Product Code. 10376244 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10376244 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10376244 Supplier Thermo Scientific Acros Supplier No. 206840050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 22661-87-6
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C2H7N3·HCl
Linear Formula CH3NHC(=NH)NH2·HCl
MDL Number MFCD00012576
Quantity 5 g
Beilstein 04, II, 571
Merck Index 10, 5953
Synonym 1-methylguanidine hydrochloride, methylguanidine hydrochloride, methylguanidine hcl, n-methylguanidine hydrochloride, 1-methylguanidine hcl, guanidine, methyl-, hydrochloride, 2-methylguanidine hydrochloride, 1-methylguanidine hydrochloride 1:x, guanidine, methyl-, monohydrochloride, methyl guanidine hcl
InChI Key VJQCNCOGZPSOQZ-UHFFFAOYSA-N
SMILES CN=C(N)N.Cl
IUPAC Name 2-methylguanidine;hydrochloride
Molecular Weight (g/mol) 109.56
PubChem CID 146724
Formula Weight 109.56
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material Methylguanidine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Methylguanidine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.