missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylhydrazine sulfate, 98%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Beskrivelse
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| Melting Point | 141°C to 143°C |
| Quantity | 25 g |
| UN Number | UN2811 |
| Beilstein | 3693575 |
| Merck Index | 14,6086 |
| Formula Weight | 144.15 |
| Percent Purity | 98% |
| Chemical Name or Material | Methylhydrazine sulfate |
Sikkerhed og håndtering
Produktidentifikator
- Methylhydrazine sulfate
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- Carcinogenicity Category 2
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H330-Fatal if inhaled.
- H351-Suspected of causing cancer.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Methylhydrazine sulfate
RUO – Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion