missing translation for 'onlineSavingsMsg'
Learn More

Methylmagnesium chloride, 3M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 11336049
Change view
Click to view available options
Quantity:
0.25 mol
11 mol
Unit Size:
0.25mol
1mol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336049 0.25 mol 0.25mol
11346049 11 mol 1mol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336049 Supplier Thermo Scientific Alfa Aesar Supplier No. 042101.JS
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 676-58-4 | CH3ClMg | 74.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 676-58-4
Molecular Formula CH3ClMg
Molecular Weight (g/mol) 74.79
MDL Number MFCD00000468
InChI Key RQNMYNYHBQQZSP-UHFFFAOYSA-M
Synonym methylmagnesium chloride, chloromethylmagnesium, chloro methyl magnesium, magnesium, chloromethyl, memgcl, methylmagnesiumchloride, methyl magnesium chloride, unii-m5e1132g4w, ch3mgcl, qmabhcaih@
PubChem CID 12670
IUPAC Name magnesium;carbanide;chloride
SMILES C[Mg]Cl

Specifications

Color Brown to Green
Physical Form Liquid
Quantity 0.25 mol
Linear Formula CH3MgCl
UN Number UN3399
Beilstein 3587199
Flash Point -17° (1°F)
Solubility Information Miscible with diethyl ether and tetrahydrofuran.
Formula Weight 74.81
Concentration or Composition (by Analyte or Components) 3M in THF
Odor Ether-like
Packaging ChemSeal™ bottles
Sensitivity Air & Moisture Sensitive
Density 1.013g/mL
Chemical Name or Material Methylmagnesium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.