missing translation for 'onlineSavingsMsg'
Learn More

Methylmalonic acid, MedChemExpress

Product Code. 16450999
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16450999 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16450999 Supplier Medchem Express Supplier No. HY103395
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Methylmalonic acid (Methylmalonate) is an indicator of Vitamin B-12 deficiency in cancer.

Methylmalonic acid (Methylmalonate) is an indicator of Vitamin B-12 deficiency in cancer.

Specifications

Chemical Name or Material Methylmalonic acid
CAS 516-05-2
Color White
Purity Grade Notes Research
Molecular Formula C4H6O4
Quantity 1 g
Synonym Methylpropanedioic acid Methylmalonate
Solubility Information DMSO : 100 mg/mL (846.81 mM; Need ultrasonic) ∣H2O : 100 mg/mL (846.81 mM; Need ultrasonic)
SMILES O=C(O)C(C)C(O)=O
Molecular Weight (g/mol) 118.09
Formula Weight 118.09
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.