Learn More
Methylmercury(II) chloride, standard solution in H{2}O, Methylmercury(II) chloride 1000ppm by AA
Description
Methylmercury(II) chloride is used as a precursor for the preparation of methyl mercury acetate. It is used as a specific reagent (electron stain) for sulfhydryl groups in biological materials like protein as well as demonstration in electron microscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 25 mL |
| Beilstein | 3600218 |
| Solubility Information | Miscible with lipid. |
| Formula Weight | 251.08 |
| Appearance | Clear |
| Chemical Name or Material | Methylmercury(II) chloride |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.