missing translation for 'onlineSavingsMsg'
Learn More

Methylparaben Sodium Salt, TRC

Product Code. 30008537
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008537 100 g 100g
30007364 25 g 25g
17962875 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008537 Supplier Toronto Research Chemicals Supplier No. TRCM338663100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5026-62-0
Molecular Formula C8 H7 O3 . Na
Molecular Weight (g/mol) 174.13
Synonym Benzoic acid, 4-hydroxy-, methyl ester, sodium salt (9CI), Benzoic acid, p-hydroxy-, methyl ester, sodium salt (6CI,8CI), Sodium, (p-carboxyphenoxy)-, methyl ester (7CI), 4-Hydroxybenzoic acid methyl ester sodium salt, Bonomold OMNa, Methyl 4-hydroxybenzoate sodium salt, Methyl p-hydroxybenzoate sodium salt, Methylparaben sodium, Preserval MS, Sodium 4-(methoxycarbonyl)phenoxide, Sodium 4-carbomethoxyphenolate, Sodium methyl 4-hydroxybenzoate, Sodium methyl p-hydroxybenzoate, Sodium methyl para-hydroxybenzoate, Sodium methylparaben, Sodium nipagin, Sodium p-methoxycarbonylphenolate, Sodium p-methoxycarbonylphenoxide, Solparol, p-Hydroxybenzoic acid methyl ester sodium salt
IUPAC Name sodium;4-methoxycarbonylphenolate
SMILES [Na+].COC(=O)c1ccc([O-])cc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H8O3.Na/c1-11-8(10)6-2-4-7(9)5-3-6;/h2-5,9H,1H3;/q;+1/p-1
Formula Weight 174.0293
Percent Purity >95%
Chemical Name or Material Sodium Methyl Parahydroxybenzoate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.