missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methylphenylphosphinic chloride, 97%

Product Code. 11493907
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493907 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11493907 Supplier Alfa Aesar™ Supplier No. A18349.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5761-97-7
Molecular Formula C7H8ClOP
Molecular Weight (g/mol) 174.56
MDL Number MFCD00013902
InChI Key BTMQENOUJJKBGT-UHFFFAOYNA-N
Synonym methylphenylphosphinic chloride, chloro methyl phosphoryl benzene, methyl phenyl phosphinoyl chloride, acmc-1arvk, methyl phenylphosphinicchloride, methylphenylphosphinyl chloride, methylphenyl-phosphinic chloride, phenyl methyl phosphinyl chloride, phosphinic chloride,methylphenyl
PubChem CID 21983
IUPAC Name [chloro(methyl)phosphoryl]benzene
SMILES CP(Cl)(=O)C1=CC=CC=C1

Specifications

Density 1.266
Boiling Point 155°C (11 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 174.57
Percent Purity 97%
Chemical Name or Material Methylphenylphosphinic chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.