missing translation for 'onlineSavingsMsg'
Learn More

Methylsuccinic acid, 99%

Product Code. 15407468
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15407468

Brand: Thermo Scientific Alfa Aesar H60967.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 498-21-5 | C5H8O4 | 132.115 g/mol

Methylsuccinic acid is used as a flux material and an important dyestuff intermediate. It is also used as an intermediate in medicine and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylsuccinic acid is used as a flux material and an important dyestuff intermediate. It is also used as an intermediate in medicine and pharmaceuticals.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature. Moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-21-5
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.115
MDL Number MFCD00002659
InChI Key WXUAQHNMJWJLTG-UHFFFAOYSA-N
Synonym 2-methylsuccinic acid, methylsuccinic acid, pyrotartaric acid, butanedioic acid, methyl, methyl succinic acid, 1,2-propanedicarboxylic acid, succinic acid, methyl, 2-methyl-butanedioic acid, butanedioic acid, 2-methyl, ccris 6068
PubChem CID 10349
IUPAC Name 2-methylbutanedioic acid
SMILES CC(CC(=O)O)C(=O)O

Specifications

Melting Point 110°C to 115°C
Linear Formula HO2CCH2CH(CH3)CO2H
Quantity 25 g
Beilstein 1722946
Solubility Information Soluble in water.
Formula Weight 132.11
Percent Purity 99%
Chemical Name or Material Methylsuccinic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.