missing translation for 'onlineSavingsMsg'
Learn More

Methylsulfamic acid, 98%, Thermo Scientific™

Product Code. 10444945 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10444945 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10444945 Supplier Thermo Scientific Acros Supplier No. 314670250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4112-03-2
Molecular Formula CH5NO3S
Molecular Weight (g/mol) 111.12
MDL Number MFCD00010870
InChI Key MYMDOKBFMTVEGE-UHFFFAOYSA-N
Synonym methylsulfamic acid, methyl sulfamic acid, methylsulamic acid, sulfamic acid, methyl, methylsulfamicacid, sulfamic acid,n-methyl, acmc-2097ek, mymdokbfmtvege-uhfffaoysa, mymdokbfmtvege-uhfffaoysa-n
PubChem CID 244258

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 189°C to 193°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3NHSO3H
Quantity 25 g
Formula Weight 111.12
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methylsulfamic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.