missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyltri-n-butylammonium bromide, 98%, Thermo Scientific™

Product Code. 11430298
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11430298

Brand: Alfa Aesar™ B23828.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37026-88-3
Molecular Formula C13H30BrN
Molecular Weight (g/mol) 280.29
MDL Number MFCD00043201
InChI Key FQUHTZCWXDFLBQ-UHFFFAOYSA-N
Synonym tributylmethylammonium bromide, methyltri-n-butylammonium bromide, 1-butanaminium, n,n-dibutyl-n-methyl-, bromide, methyltributylammonium bromide, methyl tributyl ammonium bromide, tributyl methyl azanium bromide, acmc-20ajfa, ksc493m4d, tributylmethylammonium bro-mide
PubChem CID 10062191
IUPAC Name tributyl(methyl)azanium;bromide
SMILES [Br-].CCCCC([NH3+])(CCCC)CCCC

Specifications

Melting Point 120°C to 122°C
Quantity 10 g
Beilstein 3704268
Sensitivity Hygroscopic
Formula Weight 280.29
Percent Purity 98%
Chemical Name or Material Methyltri-n-butylammonium bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.