missing translation for 'onlineSavingsMsg'
Learn More

Methyltri-n-butylammonium chloride, Aliquat 175 (75% aq. soln.)

Product Code. 11324885
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324885 500 g 500g
11304885 10 g 10g
11314885 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324885 Supplier Thermo Scientific Alfa Aesar Supplier No. L18019.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56375-79-2 | C13H30ClN | 235.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56375-79-2
Molecular Formula C13H30ClN
Molecular Weight (g/mol) 235.84
MDL Number MFCD00011847
InChI Key IPILPUZVTYHGIL-UHFFFAOYSA-M
Synonym tributylmethylammonium chloride, n,n-dibutyl-n-methylbutan-1-aminium chloride, methyltributylammonium chloride, methyl tributyl ammonium chloride, 1-butanaminium, n,n-dibutyl-n-methyl-, chloride, unii-az0b34me3u, az0b34me3u, tributyl methyl azanium chloride, 1-butanaminium, n,n-dibutyl-n-methyl-, chloride 1:1, acmc-20aker
PubChem CID 91822
IUPAC Name tributyl(methyl)azanium;chloride
SMILES CCCC[N+](C)(CCCC)CCCC.[Cl-]

Specifications

Melting Point -33°C
Density 0.964
Boiling Point 84°C to 85°C (101 mmHg)
Quantity 500 g
Beilstein 6300212
Sensitivity Hygroscopic
Formula Weight 235.84
Concentration or Composition (by Analyte or Components) 75% aq. soln.
Chemical Name or Material Methyltri-n-butylammonium chloride, Aliquat 175

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.