missing translation for 'onlineSavingsMsg'
Learn More

Methyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals

Product Code. 11955911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11955911 100 mL AcroSeal™ Glass Bottle 100mL
11965911 1 L AcroSeal™ Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11955911 Supplier Thermo Scientific Acros Supplier No. 432641000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-55-3
Molecular Formula C4H12O3Si
Molecular Weight (g/mol) 136.22
InChI Key BFXIKLCIZHOAAZ-UHFFFAOYSA-N
Synonym methyltrimethoxysilane, trimethoxy methyl silane, silane, trimethoxymethyl, union carbide a-163, silane, methyltrimethoxy, methyl trimethoxysilane, silane a-163, dynasylan mtms, unii-0hi0d71mci, methyl-trimethoxysilane
PubChem CID 14456
IUPAC Name trimethoxy(methyl)silane
SMILES CO[Si](C)(OC)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -50°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 102°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3Si(OCH3)3
Refractive Index 1.3695 to 1.3715
Quantity 100 mL
Specific Gravity 0.95
Solubility Information Solubility in water: decomposes
Viscosity 0.5 mPa.s (20°C)
Formula Weight 136.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyltrimethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyltrimethoxysilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Methyltrimethoxysilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.