missing translation for 'onlineSavingsMsg'
Learn More

Methyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals

Product Code. 11965911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
missing translation for 'unitSize':
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantity Packaging missing translation for 'unitSize'
11965911 1 L AcroSeal™ Glass Bottle 1 L
11955911 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 11965911 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 432640010
ChemsChrom Mega Deals
Use promo code 27993
Afficher toutes les promotions
 Restricted Item
Estimated Shipment: 20-11-2026
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 1185-55-3 | Molecular Formula: C4H12O3Si | Molecular Weight: 136.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1185-55-3
Molecular Formula C4H12O3Si
Molecular Weight (g/mol) 136.22
InChI Key BFXIKLCIZHOAAZ-UHFFFAOYSA-N
Synonym methyltrimethoxysilane, trimethoxy methyl silane, silane, trimethoxymethyl, union carbide a-163, silane, methyltrimethoxy, methyl trimethoxysilane, silane a-163, dynasylan mtms, unii-0hi0d71mci, methyl-trimethoxysilane
PubChem CID 14456
IUPAC Name trimethoxy(methyl)silane
SMILES CO[Si](C)(OC)OC

Spécification

CAS Min % 96.0
CAS Max % 100.0
Melting Point -50°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 102°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3Si(OCH3)3
Refractive Index 1.3695 to 1.3715
Quantity 1 L
Specific Gravity 0.95
Solubility Information Solubility in water: decomposes
Viscosity 0.5 mPa.s (20°C)
Formula Weight 136.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyltrimethoxysilane
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • Methyltrimethoxysilane
Mention d'avertissement
  • Danger
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Informations supplémentaires
  • MIXTURE LIST-Contain: Methyltrimethoxysilane

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.