missing translation for 'onlineSavingsMsg'
Learn More

Methyltrimethoxysilane, 97%

Product Code. 10472431 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10472431 250 mL Glass bottle 250mL
10296240 5 mL Glass bottle 5mL
10338520 50 mL Glass bottle 50mL
10351564 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10472431 Supplier Thermo Scientific Acros Supplier No. 250542500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1185-55-3 | C4H12O3Si | 136.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-55-3
Molecular Formula C4H12O3Si
Molecular Weight (g/mol) 136.22
MDL Number MFCD00008342
InChI Key BFXIKLCIZHOAAZ-UHFFFAOYSA-N
Synonym methyltrimethoxysilane, trimethoxy methyl silane, silane, trimethoxymethyl, union carbide a-163, silane, methyltrimethoxy, methyl trimethoxysilane, silane a-163, dynasylan mtms, unii-0hi0d71mci, methyl-trimethoxysilane
PubChem CID 14456
IUPAC Name trimethoxy(methyl)silane
SMILES CO[Si](C)(OC)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -50°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 102°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3Si(OCH3)3
Refractive Index 1.3695 to 1.3715
Quantity 250 mL
Beilstein 04, 4203
Specific Gravity 0.95
Solubility Information Solubility in water: decomposes
Viscosity 0.5 mPa.s (20°C)
Formula Weight 136.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyltrimethoxysilane, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.