missing translation for 'onlineSavingsMsg'
Learn More

Methyltrioxorhenium (VII), 98%

Product Code. 10390784 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
100mg
2.5g
500mg
This item is not returnable. View return policy

Product Code. 10390784

Brand: Thermo Scientific Acros 346960025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 70197-13-6 | CH3O3Re | 249.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70197-13-6
Molecular Formula CH3O3Re
Molecular Weight (g/mol) 249.23
MDL Number MFCD00192332
InChI Key VZSXFJPZOCRDPW-UHFFFAOYSA-N
Synonym methyltrioxorhenium vii, methyltrioxorhenium, methylrhenium trioxide, unii-883d8rdd5q, methylrhenium vii trioxide, mto, methyltrioxorhenium mi, t-4-methyltrioxorhenium, methylrhenium trioxide vii
PubChem CID 2734010
IUPAC Name carbanide;trioxorhenium
SMILES [CH3-].O=[Re](=O)=O

Specifications

Melting Point 111.0°C
Color Blue-Gray to White
CAS Min % 97.5
CAS Max % 100.0
Physical Form Crystals
Quantity 2.5 g
Assay Percent Range 97.5% min. (GC)
Merck Index 15, 6206
Solubility Information Solubility in water: very soluble in h2o
Formula Weight 249.23
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Methyltrioxorhenium (VII)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.