missing translation for 'onlineSavingsMsg'
Learn More

Methylviolettlösung for PANPEHA indicator paper, Solstice

Product Code. 17244546
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17244546 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17244546 Supplier Fluka Supplier No. 310695L
  • ONLINE EXCLUSIVE PRICE
    1455.00 €
    1312.00 € / 5 L
    Save 143.00 €
    9% Off
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material Methylviolettlösung
Melting Point -115°C (The physical data is that of the main component)
Boiling Point 78°C to 79°C (1013 hPa) (The physical data is that of the main component)
CAS 78-93-3, 64-17-5, 548-62-9
UN Number UN1170
Quantity 5 L
compliance-icons compliance-icons
Product Identifier
  • Methylviolettlo
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: butanone, ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.