missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Metricel™ Black PES Membrane Disc Filters

Contrasting membrane for microbiological analysis. Dark background provides excellent contrast for counting opaque colonies in microbiology labs.

Brand:  Cytiva (formerly Pall Lab) 66588

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 17154581

  • 158.00 € / Pack of 100
Estimated Shipment: 27-06-2024
to see stock.
Add to basket

Description

Description

  • Exclusive dot grid pattern does not enhance or inhibit colony growth certified for use in the Membrane Filter (MF) Technique
  • For research only
Specifications

Specifications

Black
0.8 μm
100/Pk.
Gridded
Hydrophilic Modified PES
Non-sterile
147 μm
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers