missing translation for 'onlineSavingsMsg'
Learn More

Metro™ HD Super Stainless Dunnage Shelves Accessory, Donut Bumper

Product Code. 15926791
Change view
Click to view available options
Material:
Plastic
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15926791 Plastic Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15926791 Supplier Metro™ Supplier No. 9992H
  • ONLINE EXCLUSIVE PRICE
    21.45 €
    16.96 € / Pack of 1
    Save 4.49 €
    20% Off
Estimated Shipment: 19-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with HD Super shelving units

  • Fits 4.1cm (1.625 in.) posts only

Specifications

Product Type Donut Bumper
Material Plastic
For Use With (Equipment) Fits 4.1cm posts only
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.