Learn More
Metro™ HD Super Stainless Dunnage Shelves Accessory, Post Clamp
Product Code. 15956791
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. Material Unit Size
15956791 Stainless Steel Each
1 options
Product Code. 15956791 Supplier Metro™ Supplier No. 9994HZ
  • ONLINE EXCLUSIVE PRICE
    20.40 €
    16.96 € / Pack of 1
    Save 3.44 €
    16% Off
Estimated Shipment: 05-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with HD Super shelving units
  • Stainless steel
  • Used to join posts together for extra stability
Specifications
Product Type Post Clamp
For Use With (Application) Used to join posts together for extra stability
For Use With (Equipment) HD Super™ shelving unit
Material Stainless Steel