missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ MetroMax i™ Lab Worktable Accessory, Drawer
The Metro MetroMax i™ Lab Worktable Drawer comes in a choice of sizes to provide extra room and functionality in your storage setup.
Brand: Metro™ LTD6
Additional Details : Weight : 3.99200kg
Descripción
- Drawers fit 30, 36, 48 and 60 in. (76, 91.4, 121.9, and 152.4cm) table lengths
- Locking feature available
Especificaciones
Drawer | |
6 in. | |
15.2 cm | |
For 30, 36, 48 and 60 in. lab worktables |