missing translation for 'onlineSavingsMsg'
Learn More

Metro™ MetroMax Q™ Posts

Product Code. 15947161
Change view
Click to view available options
Style:
Mobile
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Style Unit Size
15947161 Mobile Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15947161 Supplier Metro™ Supplier No. MQ74UPE
  • ONLINE EXCLUSIVE PRICE
    59.15 €
    51.60 € / Pack of 1
    Save 7.55 €
    12% Off
Estimated Shipment: 19-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Epoxy-coated posts have Microban™ antimicrobial product protection.

  • Stationary posts include an adjustable leveling foot to compensate for uneven floors.
  • Leveling foot can be adjusted 1 in. (25 mm).

Specifications

Product Type Stationary Shelving Post
For Use With (Equipment) Metro™ MetroMax Q™ shelving
Height (English) 74 in.
Height (Metric) 188 cm
Style Mobile
Material Epoxy-coated Steel, Polymer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.