missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Standard-Duty Mobile Security Units, Stainless Steel
Secure the most important items in your lab. Metro Standard-Duty Mobile Security Units provide protection of valuable materials and sensitive items from loss or pilferage. Available in a sleek, easy-to-clean stainless steel finish.
Brand: Metro™ SEC66S-SD
Requires : Casters must be ordered separately. Contact your Fisher Scientific™ Safety Customer Service Represenative to select and order the proper caster for your application, load requirements, and operating environment.
Description
Mobile security units provide protection for valuables and sensitive materials from loss or pilferage. Type 304 Stainless provides superior corrosion resistance. Heavy gauge open wire construction keeps contents visible at all times, making it easy to check inventory. Choice of casters to match your requirements.
- Patented, ergonomic, 1/4-turn door handle
- Double doors open 270° and can be secured along the sides of the unit
- Fixed top and bottom Super Erecta shelves
- Optional intermediate shelves can be postitioned in 1 in. increments along entire height of post
Order Info
Shipped knocked down; assemble easily. Super Adjustable/Super Erecta™ intermediate shelves and casters sold separately.
Specifications
Mobile Security Cart | |
62 in. | |
65 in. | |
Epoxy | |
215 lb. | |
85 cm |
65 x 33.5 x 62 in. (165.1 x 85 x 157.5 cm) | |
157.5 cm | |
165.1 cm | |
89 kg | |
33.5 in. |