missing translation for 'onlineSavingsMsg'
Learn More

Metronidazole, 99%

Product Code. 15426738
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426738 25 g 25g
15436738 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15426738 Supplier Thermo Scientific Alfa Aesar Supplier No. H60258.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 443-48-1 | C6H9N3O3 | 171.156 g/mol

Metronidazole is a limited spectrum antibiotic that actively deters the growth of protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria. The first use of this antibiotic was to suppress protozoans such as Entamoeba histolytica, Giardia lamblia and Trichomonas vaginalis. It has been used to repress the growth of gram-negative anaerobes belonging to the Bacteroides and Fusobacterium, and gram-positive anaerobes such aspeptostreptococcus and Clostridia. The advantages of this antibiotic is that it affects a high percentage of gram-negative bacteria and has greater tissue penetration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metronidazole is a limited spectrum antibiotic that actively deters the growth of protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria. The first use of this antibiotic was to suppress protozoans such as Entamoeba histolytica, Giardia lamblia and Trichomonas vaginalis. It has been used to repress the growth of gram-negative anaerobes belonging to the Bacteroides and Fusobacterium, and gram-positive anaerobes such aspeptostreptococcus and Clostridia. The advantages of this antibiotic is that it affects a high percentage of gram-negative bacteria and has greater tissue penetration.

Solubility
Soluble in water (10 mg/ml at 20°C), ethanol, methanol, chloroform, and dimethyl sulfoxide.

Notes
Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443-48-1
Molecular Formula C6H9N3O3
Molecular Weight (g/mol) 171.156
MDL Number MFCD00009750
InChI Key VAOCPAMSLUNLGC-UHFFFAOYSA-N
Synonym metronidazole, metronidazol, flagyl, 2-methyl-5-nitroimidazole-1-ethanol, anagiardil, trichazol, gineflavir, meronidal, metronidaz, novonidazol
PubChem CID 4173
ChEBI CHEBI:6909
IUPAC Name 2-(2-methyl-5-nitroimidazol-1-yl)ethanol
SMILES CC1=NC=C(N1CCO)[N+](=O)[O-]

Specifications

Melting Point 159°C to 161°C
Quantity 25 g
Beilstein 611683
Solubility Information Soluble in water (10mg/ml at 20°C),ethanol,methanol,chloroform,and dimethyl sulfoxide.
Formula Weight 171.16
Percent Purity 99%
Chemical Name or Material Metronidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.