missing translation for 'onlineSavingsMsg'
Learn More

Metronidazole, TRC

Product Code. 30004632
Change view
Click to view available options
Quantity:
50 mg
1 g
25 g
Unit Size:
1g
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004632 25 g 25g
17911245 1 g 1g
17904905 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004632 Supplier Toronto Research Chemicals Supplier No. TRCM33888025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443-48-1
Molecular Formula C6 H9 N3 O3
Molecular Weight (g/mol) 171.15
Synonym Metronidazole, Metronidazole Benzoate Imp. A (EP), 2-Methyl-5-nitro-1H-imidazole-1-ethanol, 1-(β-Hydroxyethyl)-2-methyl-5-nitroimidazole, 1-(2-Hydroxyethyl)-2-methyl-5-nitroimidazole, 1-Hydroxyethyl-2-methyl-5-nitroimidazole, 2-Methyl-5-nitroimidazole-1-ethanol, Anagiardil, Arilin, Bayer 5360, Cimetrol 500LPCI, Clont, Deflamon, Efloran, Elyzol, Entizol, Flagesol, Flagil, Flagyl, Fossyol, Gineflavir, Klion, Klont, Medazol, Metro Cream & Gel, MetroGel, Metrogyl, Metrolag, Metrolyl, Metron, Metronidazol, Metropol, Metrotop, Mexibol, NSC 50364, NSC 69587, Nidazole, Noritate, Orvagil, Protostat, RP 8823, Rathimed, Sanatrichom, Satric, Takimetol, Trichazol, Tricho Cordes, Tricho-Gynaedron, Trichobrol, Trichocide, Trichomol, Trichopal, Tricocet, Trikhopol, Trivazol, Vagilen, Vagimid, Zadstat
IUPAC Name 2-(2-methyl-5-nitroimidazol-1-yl)ethanol
SMILES Cc1ncc([N+](=O)[O-])n1CCO

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H9N3O3/c1-5-7-4-6(9(11)12)8(5)2-3-10/h4,10H,2-3H2,1H3
Formula Weight 171.0644
Percent Purity >95%
Chemical Name or Material Metronidazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.