Description
- Integrable and programmable antistatic device
- Eliminates electrostatic charges on sample containers
- Includes ionizer assembly and conductive glass door.
Specifications
Specifications
| For Use With (Equipment) | Excellence Plus Analytical and Micro Balance |
| For Use With (Application) | Eliminates electrostatic charges on sample containers |
| Type | Compact Anti-Static Kit |