missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Dairy pH Electrode

Product Code. 11987831
Change view
Click to view available options
Connector Types:
S7
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
11987831 S7 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11987831 Supplier METTLER TOLEDO™ Supplier No. 59904591
  • ONLINE EXCLUSIVE PRICE
    464.00 €
    391.30 € / Pack of 1
    Save 72.70 €
    15% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A pH electrode featuring a triple ceramic junction, designed for protein containing samples such as dairy products, milk and cream.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Milk and cream
  • Protein containing samples
  • Emulsion

Accessories & Care

  • Triple ceramic junction
  • A41 membrane glass
  • Refillable FRISCOLYT-B™ (51343185): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Description pH electrode InLab Dairy
Connector Types S7
Junction Triple/Ceramic
Material Glass
pH Range 0 to 12
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode
Filling Solution Type FRISCOLYT-B
For Use With (Application) Used to puncture soft media or in aqueous samples
For Use With (Equipment) S7 to BNC (30281915)
Internal Reference ARGENTHAL
Membrane A41 Glass
Model InLab™ Dairy
Storage Requirements 3mol/L KCl
Temperature (English) Operating 0°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 4.7 in.
Length (Metric) 120 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.