missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Mono Half-Cell pH Measuring Electrodes

Product Code. 10202903
Change view
Click to view available options
Temperature (Metric) Operating:
0°C to 100°C
0°C to 130°C
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature (Metric) Operating Unit Size
10202903 0°C to 100°C Each
10496313 0°C to 130°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10202903 Supplier METTLER TOLEDO™ Supplier No. 51343195
  • ONLINE EXCLUSIVE PRICE
    191.00 €
    182.99 € / Pack of 1
    Save 8.01 €
    4% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A half-cell pH measuring electrode featuring a glass body that is suited for high temperature and high pH samples in which the lifetime of the electrode is significantly shorter than that of a standard reference electrode.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • High temperature samples
  • High pH samples
  • Corrosive acids and bases
  • Drinking water

Accessories & Care

  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Connector Types S7 to BNC (30281915)
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Half-Cell Measuring Electrode; HA Membrane Glass
For Use With (Application) Electrolyte hose coupling, Electrolyte bridge
Model InLab™ Mono
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 4.7 in.
Length (Metric) 120 mm
Diameter (English) Body 0.47 in.
Diameter (Metric) Body 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.