missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Surface pH Electrodes

A pH electrode featuring a flat, rugged plastic membrane for measuring samples with flat surfaces.

Brand:  METTLER TOLEDO™ 51343157

303.58 EUR valid until 2023-12-31
Use promo code "18562" to get your promotional price.



Code : 59

Additional Details : Weight : 0.12600kg

 View more versions of this product

Product Code. 10049102

  • 325.00 € / Each

In Stock
to see stock.
Add to basket

Description

Description

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Agar plates
  • Drop size samples
  • Paper
  • Skin
  • Leather
  • Suspended solids
  • Textiles and Prints

Accessories & Care

  • Ceramic ring junction
  • Polysulfone membrane
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications

Specifications

S7
Polysulfone
0°C to 50°C
Saturated AgCl
S7 to BNC (30281915)
Ag/AgCl
InLab™ Surface
3mol/L KCl
0°C to 50°C
110 mm
12 mm
Ceramic
1 to 11
Combination Electrode
For measuring samples such as Skin, Leather, Paper and Agar
Flat membrane
LoT Glass
<20 sec.
32°F to 122°F
4.33 in.
0.47 in.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers