missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Viscous pH Electrodes

Artikelnummer. 11977841
Ansicht ändern
Click to view available options
Connector Types:
Multi-Pin
S7
Packungsgröße:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Connector Types Packungsgröße
11977841 Multi-Pin Each
12570137 S7 Each
10335303 S7 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11977841 Lieferant METTLER TOLEDO™ Lieferanten-Nr. 51343151
  • ONLINE EXCLUSIVE PRICE
    856.00 €
    750.00 € / Pack of 1
    Save 106.00 €
    12% Off
Alle Werbeaktionen anzeigen
In Stock
Zum Warenkorb hinzufügen
Edge
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

A glass pH electrode with an integrated temperature sensor featuring the SteadyForce™ reference system, which is pressurized to prevent the junction from clogging.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Cosmetics
  • Gels, soaps, shampoos
  • Resins
  • Paints
  • Varnishes and glues
  • Colorants and dyes
  • Oily samples
  • TRIS compatible

Accessories & Care

  • Ceramic Junction and non-refillable - no additional electrolyte required
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Spezifikation

Description pH Electrode InLab Viscous Pro-ISM
Connector Types Multi-Pin
Junction Ceramic
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 130°C
Type Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology
Filling Solution Type FRISCOLYT-C
For Use With (Application) Highly viscous, highly glutinous samples
For Use With (Equipment) Multi-Pin to BNC/RCA (30281896)
Internal Reference SteadyForce
Membrane HA Glass
Model InLab™ Viscous Pro-ISM
Probe Integrated NTC 30 kω Temp Probe
Storage Requirements 3 mol/L KCl
Temperature (English) Operating 32°F to 266°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 1.57 in.
Length (Metric) 40 mm
Diameter (English) 0.5 in.
Diameter (Metric) 12 mm
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.