missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Reference Electrodes

Product Code. 10018622 Shop All Mettler Toledo Products
Click to view available options
Junction:
Ceramic
PTFE Sleeve
Triple/Ceramic
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10018622

Brand: METTLER TOLEDO™ 51343191

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

A reference electrode to be paired with half-cell measurement electrodes.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • For use with half-cell measuring electrodes

Accessories & Care

  • S7 connector type: Cable S7 to 2 mm Banana (30281923) required
  • InLab Reference: ceramic junction
  • InLab Reference Plus: moveable PTFE sleeve junction
  • InLab Reference Flow: triple ceramic junction
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Connector Types S7
Junction PTFE Sleeve
Material Glass
Temperature (Metric) Operating 0°C to 60°C
Type Half-Cell Reference Electrode
Filling Solution Type 3mol/L KCl
For Use With (Application) For routine applications and measurements in Ion-selective half cells
Internal Reference AAg/AgCl
Model InLab™ Reference Plus
Reference Electrolyte Gel
Temperature (English) Operating 32°F to 140°F
Temperature (Metric) Range 0°C to 60°C
Length (English) 4.72 in.
Length (Metric) 12 cm
Diameter (English) Body 0.47 in.
Diameter (Metric) Body 1.2 cm
Show More Show Less
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt