missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ ISA Solutions

Product Code. 11997511
Change view
Click to view available options
Type:
ISA Solution
TISAB II with CDTA
TISAB III with CDTA
For Use With (Equipment):
Calcium Electrode
Fluoride Electrode
Nitrate Electrode
Potassium Electrode
Unit Size:
3790mL
475mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Unit Size
11997511 TISAB II with CDTA Fluoride Electrode 3790mL
11957511 ISA Solution Calcium Electrode 475mL
11977511 ISA Solution Nitrate Electrode 475mL
12797421 ISA Solution Potassium Electrode 475mL
11907521 TISAB III with CDTA Fluoride Electrode 475mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11997511 Supplier Mettler Toledo™ Supplier No. 51344765
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Solutions for perfectION™ combined ion selective electrodes

Solutions (ionic-strength adjustment buffers)

  • Ion Electrolyte B (for chloride, copper, iodide, silver/sulfide ISE's), 475mL
  • ISA for calcium electrode, 475mL
  • ISA for potassium electrode, 475mL
  • ISA for nitrate electrode, 475mL
  • Nitrate ISS (for suppressing interference), 475mL
  • Fluoride TISAB II with CDTA, 3790mL
  • Fluoride TISAB III with CDTA (concentrate), 475mL

TRUSTED_SUSTAINABILITY

Specifications

Description perfectION TISAB II with CDTA
For Use With (Equipment) Fluoride Electrode
Type TISAB II with CDTA
Filling Solution Type Fluoride TISAB II with CDTA
Packaging 3790 mL
Quantity 3790 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.