missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Redox ORP Electrodes

Product Code. 10786733 Shop All Mettler Toledo Products
Click to view available options
Model:
InLab Redox
InLab Redox Ag
InLab Redox Ag805
InLab Redox Au
InLab Redox Flow
InLab Redox Micro
InLab Redox Pt805
InLab Redox-L
Type:
Combination Electrode
Half Cell Electrode
Electrode:
Gold Ring Indicator
Platinum Ring Indicator
Silver Ring Indicator
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10786733

Brand: METTLER TOLEDO™ 51343200

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

An ORP/Redox electrode featuring a metal ring indicator designed for measuring oxidation/reduction potential; ideal for water and other general applications.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Measuring oxidation/reduction potential (ORP)
  • Drinking water
  • Cooling towers
  • Swimming pools
  • Potable water supplies
  • Waste water
  • General lab

Accessories & Care

  • Ceramic junction (excluding the InLab Redox Flow with movable glass sleeve junction)
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • InLab Redox Pt805 and InLab Redox Ag805: no electrolyte required and must be paired with InLab Reference (51343190)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Spécification

Connector Types S7
Junction Ceramic
Material Glass
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode
Electrode Platinum Ring Indicator
Filling Solution Type 3mol/L KCl
For Use With (Application) For measuring the oxidation/reduction power of solution
For Use With (Equipment) S7 to BNC (30281915)
Internal Reference ARGENTHAL with Ag+ trap
Model InLab Redox
Storage Requirements 3mol/L KCl
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 4.72 in.
Length (Metric) 120 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis